Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium carbonate, 99+%, for biochemistry, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
471-34-1
CCaO3
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama
10112
CHEBI:3311
calcium;carbonate
C(=O)([O-])[O-].[Ca+2]
471-34-1
CCaO3
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama
10112
CHEBI:3311
calcium;carbonate
C(=O)([O-])[O-].[Ca+2]
Assay Percent Range 99% min. (Complexometry)
Linear Formula CaCO3
Fieser 01,103; 02,57; 04,67; 05,89
Merck Index 15,1659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids
Formula Weight 100.09
Percent Purity 99+%
Grade Biochemistry
Chloride 0.005% max
Insoluble Matter 0.01% max. (in HCl)
Packaging Plastic bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in diluted acids
Sulfate 0.02% max
Melting Point 825.0°C
Quantity 500g
Chemical Name or Material Calcium carbonate
Show More Show Less

EINECSNumber : 207-439-9