Learn More
Calcium Carbonate, Light Powder, FCC, 98-100.5%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Pail
C1078, 471-34-1, CaCO3
Spectrum™ Calcium Carbonate, Light Powder, FCC is used as a leavening agent, yeast nutrient, pH regulator in food applications. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Arsenic (As) 3 mg/kg
Fluoride 5e-005
Insoluble in Acid 0.002
Lead (Pb) 3 mg/kg
Loss on Drying 0.02
Packaging Poly Pail