Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Light Powder, FCC, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
471-34-1
CCaO3
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
[Ca++].[O-]C([O-])=O
471-34-1
CCaO3
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
[Ca++].[O-]C([O-])=O

C1078, 471-34-1, CaCO3

Spectrum™ Calcium Carbonate, Light Powder, FCC is used as a leavening agent, yeast nutrient, pH regulator in food applications. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 mg/kg
Percent Purity 98-100.5%
Fluoride 0.005%
Insoluble in Acid 0.2%
Lead (Pb) 3 mg/kg
Loss on Drying 2%
Packaging Fiber Drum
Quantity 12 kg