Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Light Powder, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

471-34-1
CCaO3
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
CHEBI:3311
SMILES C(=O)([O-])[O-].[Ca+2]
IUPAC Name calcium;carbonate
Formula Weight 100.09
Physical Form Powder
Packaging Poly Pail
Quantity 2.5kg
Chemical Name or Material Calcium Carbonate
Grade FCC/USP