Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Light Powder, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
471-34-1
CCaO3
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
[Ca++].[O-]C([O-])=O
471-34-1
CCaO3
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
[Ca++].[O-]C([O-])=O

CA130, 471-34-1, CaCO3

Spectrum™ Calcium Carbonate, Light Powder, USP is used as a calcium supplement and as an antacid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 ppm
Percent Purity 98-100.5%
Fluoride 50 ppm
Insoluble in Acid 0.2%
Lead (Pb) 3 ppm
Loss on Drying 2%
Mercury (Hg) 0.5 ppm
Packaging Fiber Drum
Quantity 12 kg