Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Low in Alkalis, Powder, ACS, 99%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
471-34-1
471-34-1
Grade ACS
CAS Max % 1
Ammonium 3e-05
Barium (Ba) 0.0001
Percent Purity 0.99
Chloride 1e-05
Fluoride 1.5e-05
Heavy Metals 1e-05
Iron (Fe) 2e-05
Magnesium (Mg) 0.0001
Packaging Poly Pail
Potassium (K) 0.0001
Sodium (Na) 0.0001
Strontium (Sr) 0.001
Sulfate 5e-05
Quantity 2.5 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.