Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Powder, ACS, 99%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
471-34-1
471-34-1
Grade ACS
CAS Max % 1
Ammonium 3e-05
Barium (Ba) 0.0001
Percent Purity 0.99
Chloride 1e-05
Fluoride 1.5e-05
Heavy Metals 1e-05
Iron (Fe) 3e-05
Magnesium (Mg) 0.0002
Packaging Amber Glass Bottle
Potassium (K) 0.0001
Sodium (Na) 0.001
Strontium (Sr) 0.001
Sulfate 0.0001
Quantity 500 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.