Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Powder, Low in Alkalies, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle

Chemical Identifiers

471-34-1
CCaO3
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
CHEBI:3311
calcium;carbonate
C(=O)([O-])[O-].[Ca+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 100.09
Physical Form Powder
Packaging Poly Bottle
Quantity 2kg
Chemical Name or Material Calcium Carbonate
Grade Baker Analyzed™ ACS Reagent