Alfa Aesar™ Calcium carbonate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4723522

Quantity 100g

 View more versions of this product

Catalog No. AA4723522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 471-34-1
    Formula Weight 100.09
    MDL Number MFCD00010906
    Merck Index 14,1657
    Chemical Name or Material Calcium carbonate
    Grade Puratronic™

    Specifications

    Formula Weight 100.09
    MDL Number MFCD00010906
    TSCA TSCA
    CAS 471-34-1
    Merck Index 14,1657
    Color White
    Density 2.930g/mL
    RTECS Number FF9335000
    Physical Form Powder
    Odor Odorless
    Melting Point 800°C (dec.)
    Quantity 100g
    Chemical Name or Material Calcium carbonate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit