Learn More
Calcium Channel beta-4 Mouse anti-Human, Mouse, Rat, Clone: S10-7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527712
Catalog No. PIMA527712
Add to cart
Description
Description
This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy and juvenile myoclonic epilepsy. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
Calcium Channel beta-4
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
D4A055, O00305, Q8R0S4
CAB4, CACNLB4, EA5, EJM, Calcium channel voltage dependent beta 4 subunit
Mouse
IgG1
100 μg
-20°C
Primary
12298, 58942, 785
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
S10-7
Unconjugated
CACNB4
Liquid
Cacnb4
Synthetic peptide amino acids 458-474 of rat CavB4
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat