Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, 0.5 M Solution, Spectrum™

Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 10035-04-8,7732-18-5
Concentration 0.5 M ± 0.02 M
Packaging Poly Bottle
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.