Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride, 96%, for biochemistry, anhydrous, ACROS Organics™

Click to view available options
Quantity:
100g
500g
10043-52-4
CaCl2
110.99
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca+2]
10043-52-4
CaCl2
110.99
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca+2]
Assay Percent Range 95% min. (Argentometry)
Linear Formula CaCl2
Ammonium 0.005% max
Fieser 01,104; 04,368
Merck Index 15,1661
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
Formula Weight 110.99
Percent Purity 96%
Grade Biochemistry
Packaging Plastic bottle
Phosphate 0.001% max
Solubility Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
Sulfate 0.02% max
Boiling Point >1600.0°C
Melting Point 782.0°C
Quantity 100g
Chemical Name or Material Calcium chloride
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 233-140-8