Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride, 96%, extra pure, powder, anhydrous, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
10043-52-4
CaCl2
110.99
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca+2]
10043-52-4
CaCl2
110.99
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca+2]
Adsorbent
Assay Percent Range 95% min. (Complexometry)
Linear Formula CaCl2
Fieser 01,104; 04,368
Merck Index 15,1661
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
Barium (Ba) 200ppm max.
Formula Weight 110.99
Physical Form Powder
Percent Purity 96%
Grade Extra Pure
Heavy Metals (as Pb) 50ppm max.
Insoluble in Water 0.1% max.
Insoluble Matter 0.1% max. (in water)
Iron (Fe) 50ppm max.
Magnesium (Mg) 1000ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
Sulfate 500ppm max.
Boiling Point >1600.0°C
Color White
Melting Point 782.0°C
Quantity 25g
Chemical Name or Material Calcium chloride, 96%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-140-8

RTECSNumber : EV9800000

TSCA : TSCA