Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium chloride, ACS reagent, desiccant, ACROS Organics™
Click to view available options
Quantity:
100g
500g
Specifications
Linear Formula | CaCl2 |
Fieser | 01,104; 04,368 |
Merck Index | 15,1661 |
Solubility Information | Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol |
Formula Weight | 110.99 |
Grade | ACS Reagent |
Solubility | Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol |
Boiling Point | >1600.0°C |
Melting Point | 782.0°C |
Quantity | 500g |
Show More |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 233-140-8