Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Anhydrous, 20 Mesh and finer, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

10043-52-4
CaCl2
110.978
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca+2]

High quality chemicals for laboratory and specialized industrial use

    For desiccant use
Formula Weight 110.98
Mesh Size 20 Mesh and finer
Packaging Poly Bottle
Specific Gravity 1L = 2.15kg
Quantity 500g
Chemical Name or Material Calcium Chloride, Anhydrous
Grade Baker Analyzed™ ACS Reagent