Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Chloride, Anhydrous, 20 Mesh and finer, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Description
- For desiccant use
Specifications
Formula Weight | 110.98 |
Grade | BAKER ANALYZED™ ACS Reagent |
Mesh Size | 20 Mesh and finer |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.15kg |
Quantity | 2.5kg |
Chemical Name or Material | Calcium Chloride, Anhydrous |