Learn More
Calcium chloride, anhydrous, ACS, 96.0% min, Thermo Scientific Chemicals
Description
Typically an ionic halide often used as a dessicant,Calcium chloride is used as a drying and dehydrating agent for organic liquids and gases and for solids in desiccators. Common applications include brine for refrigeration plants and ice and dust control on roads. Use of calcium chloride and urea has been reported to significantly increase average fruit weight and ascorbic acid content in pomegranate. The strawberry fruit storability can also be improved by edible coating of calcium chloride. It is used in drying tubes in research laboratories, salt/chemical based dehumidifiers, fire-extinguishers, plastics, ceramic slipware, as flux in the Davy process for the manufacture of sodium and to increase water hardness in swimming pool. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. CaCl2-Pybox (pyridine flanked by two oxazoline groups) has been reported to be an efficient chiral catalyst for asymmetric 1,4-addition reactions of 1,3-dicarbonyl compounds with nitroalkenes, affording gamma-nitro carbonyl compounds in high enantioselectivities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Boiling Point | >1600°C |
Melting Point | 782°C |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 96.0% min. |
Merck Index | 14,1659 |
Solubility Information | Freely soluble in water,ethanol,methanol and acetic acid. Insoluble in liquid ammonia,dimethyl sulfoxide and ethyl acetate. |
Formula Weight | 110.99 |
Percent Purity | ≥96.0% |
Grade | ACS |
Show More |
Safety and Handling
Hazard Category | H319-H500 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P264b-P280i-P305+P351+P338-P312-P501c |
EINECSNumber | 233-140-8 |
RTECSNumber | EV9800000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.