Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Anhydrous, Desiccant, 20 Mesh, Approx. 95%, Spectrum™

Click to view available options
Quantity:
500 g,500 g
2.5 kg,2.5 kg
6 x 500 g,6 x 500 g
12 kg,12 kg
4 x 2.5 kg,4 x 2.5 kg
50 kg,50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10043-52-4
CaCl2
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
[Cl-].[Cl-].[Ca++]
10043-52-4
CaCl2
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
[Cl-].[Cl-].[Ca++]

C1090, 10043-52-4, CaCl2

Spectrum™ Calcium Chloride, Anhydrous, Desiccant, 20 Mesh is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and must be kept in tightly sealed, air tight containers. Calcium chloride has many uses but its main use is as a desiccant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Quantity 500 g,500 g
CAS Max % 100%
Packaging Amber Glass Bottle
Percent Purity Approx. 95%