Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Anhydrous, Desiccant, 20 Mesh, Approx. 95%, Spectrum™ Chemical
SDP

Catalog Number 18601235
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10043-52-4
CaCl2
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
[Cl-].[Cl-].[Ca++]
10043-52-4
CaCl2
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
[Cl-].[Cl-].[Ca++]

C1090, 10043-52-4, CaCl2

Spectrum™ Chemical Calcium Chloride, Anhydrous, Desiccant, 20 Mesh is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and must be kept in tightly sealed, air tight containers. Calcium chloride has many uses but its main use is as a desiccant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 100%
Quantity 4 x 2.5 kg
Packaging Poly Pail
Percent Purity ∼95%
Grade Reagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit