Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Desiccant, ACS, 96%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10043-52-4
10043-52-4
Grade ACS
CAS Max % 1
Percent Purity 0.96
Packaging Fiber Drum
Titratable Base 0.006 meq/g
Quantity 50 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.