Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Fisher Science Education™ Calcium Chloride, Dihydrate

Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade - Flakes
Description
This product is for educational use - no C of A is available.
Specifications
Color | White |
Quantity | 100 g |
Assay Percent Range | >95% w/w |
Linear Formula | CaCl2·2H2O |
Formula Weight | 147.02 |
Packaging | Lab Grade - Flakes |
Density | 0.83g/mL |
Chemical Name or Material | Calcium Chloride, Dihydrate, Dihydrate |
Safety and Handling

ShelfLife : 1 year
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title