Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride dihydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic Bottle
Plastic drum
10035-04-8
CaCl2·2H2O
147.02
10035-04-8
CaCl2·2H2O
147.02
Adsorbent
Assay Percent Range 99% min. (Complexometry)
Linear Formula CaCl2·2H2O
Fieser 01,104
Merck Index 15,1661
Solubility Information Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Barium (Ba) 50ppm max.
Formula Weight 147.02
Physical Form Crystals or Crystalline Powder
Percent Purity 99+%
Grade Analysis
Density 0.8300g/mL
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 5ppm max.
Magnesium (Mg) 50ppm max.
Packaging Plastic Bottle
Solubility Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Specific Gravity 0.83
Strontium (Sr) 1000ppm max.
Sulfate 100ppm max.
Color White
Melting Point 175.0°C
pH 4.5 to 6.5 (5% soln. at 20°C)
Quantity 2.5kg
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less

GHS Signal Word: Warning