Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, BiotechGrade, 99-103%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
10035-04-8
CaCl2H4O2
147.01
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
calcium dihydrate dichloride
O.O.[Cl-].[Cl-].[Ca++]
10035-04-8
CaCl2H4O2
147.01
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
calcium dihydrate dichloride
O.O.[Cl-].[Cl-].[Ca++]

C1086, 10035-04-8, CaCl2•2H2O

Spectrum™ Calcium Chloride, Dihydrate, BiotechGrade is a white deliquescent compound which is commonly used as a reagent in biochemistry. Biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability rely on calcium ions. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
CAS Max % 100%
Aluminum (Al) 1 μg/g
Appearance White crystals
Arsenic (As) 3 ppm
Percent Purity 99-103%
Fluoride 0.004%
Heavy Metals 0.0005%
Lead (Pb) 0.1%
Packaging Poly Bottle
pH Range 4.5- 9.2
Sulfate 0.01%
Quantity 2.5 kg
Show More Show Less