Learn More
Calcium Chloride, Dihydrate, EP, BP, JP, USP, 99-103%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Quantity:
500 g
CA140, 10035-04-8, CaCl2•2H2O
Spectrum™ Calcium Chloride, Dihydrate, USP, EP, BP, JP is used in treatment of hypocalcemia, electrolyte depletion, and hyperkalemia. All Spectrum Chemical USP, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Arsenic (As) 2 ppm
Iron (Fe) 10 ppm
Packaging Poly Pail
Sulfate 0.00024