Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2, ACROS Organics™

Click to view available options
Quantity:
2.5kg
10kg
Packaging:
Plastic Bottle
Plastic drum
10035-04-8
CaCl2·2H2O
MFCD00149613
10035-04-8
CaCl2·2H2O
MFCD00149613
Adsorbent
Linear Formula CaCl2·2H2O
Fieser 01,104
Merck Index 15, 1661
Formula Weight 147.02
Physical Form Flakes
Percent Purity 77 to 80% (CaCl2)
Grade Extra Pure
Density 0.8300g/mL
Packaging Plastic Bottle
Solubility Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Specific Gravity 0.83
Color Gray-White to White
Melting Point 175.0°C
Quantity 2.5kg
Chemical Name or Material Calcium chloride dihydrate, For drying pursposes
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning