Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, FCC, 99-107%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10035-04-8
CaCl2H4O2
147.01
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
calcium dihydrate dichloride
O.O.[Cl-].[Cl-].[Ca++]
10035-04-8
CaCl2H4O2
147.01
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
calcium dihydrate dichloride
O.O.[Cl-].[Cl-].[Ca++]

C1099, 10035-04-8, CaCl2•2H2O

Spectrum™ Calcium Chloride, Dihydrate, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 mg/kg
Percent Purity 99-107%
Fluoride 0.004%
Lead (Pb) 5 mg/kg
Packaging Poly Pail
Quantity 2.5 kg