Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, Reagent, ACS, 99-105%, Spectrum™

Click to view available options
Quantity:
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10035-04-8
CaCl2H4O2
147.01
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
calcium dihydrate dichloride
O.O.[Cl-].[Cl-].[Ca++]
10035-04-8
CaCl2H4O2
147.01
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
calcium dihydrate dichloride
O.O.[Cl-].[Cl-].[Ca++]

C1096, 10035-04-8, CaCl2•2H2O

Spectrum™ Calcium Chloride, Dihydrate, Reagent, ACS is a analytical grade chemical that is packaged in laboratory size bottles to production drum quantities. As an ACS grade Reagent, Spectrum Chemical manufactured Calcium Chloride is used as the quality standard against which other Calcium Chloride substances are graded and has met the toughest regulatory standards for quality and pureness.
CAS Max % 1
Percent Purity 99 to 105%
Packaging Poly Bottle
pH Range 4.5- 8.5
Strontium (Sr) 0.001
Quantity 500 g