Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Chloride, Dihydrate, Reagent, ACS, 99-105%, Spectrum™ Chemical
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Calcium Chloride, Dihydrate, Reagent, ACS is a analytical grade chemical that is packaged in laboratory size bottles to production drum quantities. As an ACS grade Reagent, Spectrum Chemical manufactured Calcium Chloride is used as the quality standard against which other Calcium Chloride substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
pH | 4.5- 8.5 |
CAS Max % | 100% |
Quantity | 4 x 2.5 kg |
Percent Purity | 99 to 105% |
Grade | ACS |
Packaging | Poly Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction