Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Calcium chloride hydrate, Puratronic™, 99.9965% (metals basis)

Click to view available options
Quantity:
2g
10g
50g
250g
22691-02-7
CaCl2H12O6
219.068
MFCD00149616
QHFQAJHNDKBRBO-UHFFFAOYSA-L
calcium chloride hexahydrate,antarcticite,acmc-20alis,calcium hexahydrate dichloride,calcium chloride-water 1/6,calcium chloride hydrate, puratronic,calcium chloride, hydrated,calcium chloride hydrate trace metals basis 1g
6093252
calcium;dichloride;hexahydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
22691-02-7
CaCl2H12O6
219.068
MFCD00149616
QHFQAJHNDKBRBO-UHFFFAOYSA-L
calcium chloride hexahydrate,antarcticite,acmc-20alis,calcium hexahydrate dichloride,calcium chloride-water 1/6,calcium chloride hydrate, puratronic,calcium chloride, hydrated,calcium chloride hydrate trace metals basis 1g
6093252
calcium;dichloride;hexahydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
Merck Index 14,1659
Solubility Information Soluble in water (5360mg/ml at 20°C),and alcohol.
Formula Weight 110.99 (Anhydrous)
Physical Form Crystalline Aggregates
Percent Purity 99.9965%
Grade Puratronic™
Density 1.71 g/mL
Odor Odorless
Sensitivity Hygroscopic
Quantity 250g
Chemical Name or Material Calcium chloride hydrate
Assay (metals basis)
Show More Show Less

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 607-129-7

TSCA : Yes

Recommended Storage : Ambient temperatures