Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Pellets, Purified, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
10043-52-4
CaCl2
110.978
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca+2]
10043-52-4
CaCl2
110.978
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride,calcium dichloride,calcium chloride anhydrous,calcium ii chloride,calciumchloride,caloride,liquical,calcium chloride pellets,isocal,jarcal
5284359
CHEBI:3312
calcium;dichloride
[Cl-].[Cl-].[Ca+2]

For Desiccant Use

Formula Weight 110.98
Physical Form Pellet
Grade Purified
Packaging Poly Bottle
Specific Gravity 1L = 2.15kg
Quantity 500g
Chemical Name or Material Calcium Chloride