Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Pellets, Purified, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

10043-52-4
CaCl2
110.978
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
CHEBI:3312

For Desiccant Use

CAS 10043-52-4
Molecular Formula CaCl2
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Ca+2]
IUPAC Name calcium;dichloride
Molecular Weight (g/mol) 110.978
PubChem CID 5284359
ChEBI CHEBI:3312
Formula Weight 110.98
Physical Form Pellet
Packaging Poly Bottle
Specific Gravity 1L = 2.15kg
Quantity 2.5kg
Chemical Name or Material Calcium Chloride
Grade Purified
Show More Show Less