Learn More
Calcium Citrate, Tetrahydrate, Granular, FCC, 97.5-100.5%, Spectrum™
Click to view available options
Packaging:
Poly Bottle
Poly Pail
C1258, 5785-44-4, C12H10Ca3O14•4H2O
Spectrum™ Calcium Citrate, Tetrahydrate, Granular, FCC is an odorless white powder and the calcium salt of citric acid and is commonly used as a preservative and flavorant food additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Lead (Pb) 2 mg/kg
Loss on Drying 10.0-14%
Packaging Poly Pail