Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Citrate, Tetrahydrate, Powder, FCC, 97.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
56.7 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5785-44-4
C12H18Ca3O18
570.49
LNIZKKFWMDARJV-UHFFFAOYSA-H
tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
5785-44-4
C12H18Ca3O18
570.49
LNIZKKFWMDARJV-UHFFFAOYSA-H
tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

CA142, 5785-44-4, C12H10Ca3O14•4H2O

Spectrum™ Calcium Citrate, Tetrahydrate, Powder, FCC is an odorless white powder and the calcium salt of citric acid and is commonly used as a preservative and flavorant food additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 10.0-14%
Quantity 12 kg
Packaging Poly Pail
CAS Max % 100%
Percent Purity 97.5-100.5%