Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Calcium Deficient Diet, Pellet

Calcium signaling animal diet.

$1175.00 - $4460.00

Specifications

Type Animal Diet
Storage Requirements Room Temperature
Disposable Yes
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP290121610
View Documents
MP Biomedicals
0290121610
10 kg
Each for $1,175.00
Add to cart
 
MP290121620
View Documents
MP Biomedicals
0290121620
20 kg
Each for $2,060.00
Add to cart
 
MP290121650
View Documents
MP Biomedicals
0290121650
50 kg
Each for $4,460.00
Add to cart
 
Description

Description

Calcium deficient diet can be used to understand the calcium deficiency on bone density, osteoporosis and other calcium signaling pathways.
Specifications

Specifications

Animal Diet
Room Temperature
Yes
Non-sterile
SDS
Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit