Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Disodium EDTA, FCC, 97-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
23411-34-9
C10H16CaN2Na2O9
394.30
WJEKUTCBXSHIOX-UHFFFAOYSA-J
O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O
23411-34-9
C10H16CaN2Na2O9
394.30
WJEKUTCBXSHIOX-UHFFFAOYSA-J
O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O

E1194, 23411-34-9, C10H12CaN2Na2O8•xH2O

Spectrum™ Calcium Disodium EDTA, FCC is used as a flavor and color preservative. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 97-102%
Lead (Pb) 4 mg/kg
Packaging Poly Bottle
pH Range 6.5-7.5
Quantity 125 g