Alfa Aesar™ Calcium fluoride, 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 1099814

Quantity 25g

 View more versions of this product

Catalog No. AA1099814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-75-5
    Formula Weight 78.08
    MDL Number MFCD00010907
    Merck Index 14,1667
    Chemical Name or Material Calcium fluoride

    Specifications

    Formula Weight 78.08
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Refractive Index 1.434
    Sensitivity Hygroscopic
    MDL Number MFCD00010907
    Mesh Size −325 Mesh
    Merck Index 14,1667
    Boiling Point ≈2500°C
    Quantity 25g
    Health Hazard 1 Warning
    Density 3.18g/mL
    TSCA TSCA
    Physical Form Powder
    CAS 7789-75-5
    Melting Point 1403°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number EW1760000
    Chemical Name or Material Calcium fluoride
    Assay 99.95%, (metals basis)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit