Alfa Aesar™ Calcium fluoride, precipitated, 97%

Manufacturer: Alfa Aesar™ A101760I

Quantity 5000g

 View more versions of this product

Catalog No. AAA101760I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-75-5
    Molecular Formula CaF2
    Formula Weight 78.08
    MDL Number MFCD00010907
    Merck Index 14,1667
    Chemical Name or Material Calcium fluoride, Precipitated

    Specifications

    Formula Weight 78.08
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7789-75-5
    Quantity 5000g
    MDL Number MFCD00010907
    Merck Index 14,1667
    Boiling Point ≈2500°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number EW1760000
    TSCA TSCA
    Odor Odorless
    Molecular Formula CaF2
    Melting Point 1403°C
    Chemical Name or Material Calcium fluoride, Precipitated
    Assay 97%

    Description

    Applications
    Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.

    Solubility
    Soluble in ammonium salts. Soluble in water (slightly).

    Notes
    Store at room temperature. Separated from mineral acids. Protect from humidity and water. Incompatible with moisture and oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit