Alfa Aesar™ Calcium fluoride sputtering target, 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

Manufacturer: Alfa Aesar™ 43343KS

Quantity 1 each

 View more versions of this product

Catalog No. AA43343KS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-75-5
    Molecular Formula CaF2
    Formula Weight 78.08
    MDL Number MFCD00010907
    Chemical Name or Material Calcium fluoride sputtering target, 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick

    Specifications

    Formula Weight 78.08
    Odor Odorless
    CAS 7789-75-5
    Molecular Formula CaF2
    Quantity 1 each
    TSCA TSCA
    MDL Number MFCD00010907
    Chemical Name or Material Calcium fluoride sputtering target, 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick
    Assay 99.95%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit