Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium fluoride, White powder, 99.99% (Metals basis), Alfa Aesar™
Click to view available options
Quantity:
5g
25g
100g
Specifications
Assay Percent Range | 99.99% (Metals basis) |
Beilstein | EM>5, |
Solubility Information | Slightly soluble in acid. Insoluble in acetone. |
Formula Weight | 78.08 |
Physical Form | Powder |
Density | 3.18 |
Odor | Odorless |
Refractive Index | 1.434 |
Sensitivity | Hygroscopic |
Boiling Point | ca 2500°C |
Show More |
Safety and Handling
H315-H319-H335-H303 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.
EINECSNumber : 232-188-7
RTECSNumber : EW1760000
TSCA : Yes
Recommended Storage : Ambient temperatures