Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Calcium Free Salt Mixture, Modified
No Calcium in salt mixture.
Supplier: MP Biomedicals 0296020410
Description
- Salt mixture for multiple uses
- No calcium in mixture
- Modified to include essential trace minerals
- This composition is high in phosphate
Specifications
Not Autoclavable | |
Yes | |
Non-sterile | |
1 ft. |
Animal Diet | |
10 kg | |
Room Temperature | |
0.3048 m |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?
Provide Content Correction