Learn More
Calcium Gluceptate, Hemiheptahydrate, USP, 95-102%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
C1120, 29039-00-7, C14H26CaO16•31/2H2O
Spectrum™ Calcium Gluceptate, Hemiheptahydrate, USP is used in the management of hypocalcaemia and calcium deficiency states resulting from dietry deficiency or aging. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chloride 0.0007
Heavy Metals (as Pb) 2e-005
Loss on Drying 0.114
Packaging Fiber Drum
Sulfate 0.0005