Learn More
Calcium Gluconate, Anhydrous, FCC, 98-102%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Pail
C1113, 299-28-5, C12H22CaO14
Spectrum™ Calcium Gluconate, Anhydrous, FCC is used as a nutrient, firming agent, stabilizer, and texturizer in grain products and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Lead (Pb) 2 mg/kg
Loss on Drying 0.03
Packaging Poly Pail