Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Gluconate, Anhydrous, FCC, 98-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
299-28-5
C12H22CaO14
430.37
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
[Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
299-28-5
C12H22CaO14
430.37
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
[Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

C1113, 299-28-5, C12H22CaO14

Spectrum™ Calcium Gluconate, Anhydrous, FCC is used as a nutrient, firming agent, stabilizer, and texturizer in grain products and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 98-102%
Lead (Pb) 2 mg/kg
Loss on Drying 3%
Packaging Fiber Drum
Quantity 50 kg