Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Glycerophosphate, FCC, 98-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
27214-00-2
C3H7CaO6P
210.14
UHHRFSOMMCWGSO-UHFFFAOYSA-L
calcium 2-(phosphonatooxy)propane-1,3-diol
[Ca++].OCC(CO)OP([O-])([O-])=O
27214-00-2
C3H7CaO6P
210.14
UHHRFSOMMCWGSO-UHFFFAOYSA-L
calcium 2-(phosphonatooxy)propane-1,3-diol
[Ca++].OCC(CO)OP([O-])([O-])=O

CA147, 27214-00-2, C3H7CaO6P

Spectrum™ Calcium Glycerophosphate, FCC is used to neutralize acidic foods when consumed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 98-100.5%
Lead (Pb) 4 mg/kg
Loss on Drying 12%
Packaging Amber Glass Bottle
Quantity 500 g