Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium granules, -9 mesh, 98.8% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
50g
250g
1kg
7440-70-2
Ca
40.08
MFCD00085314,MFCD00010897
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]
7440-70-2
Ca
40.08
MFCD00085314,MFCD00010897
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]
Quantity 50g
Appearance Silvery-white granules
Density 1.54g/cm3
Assay Percent Range 98.8% (Metals basis)
Mesh Size -9 Mesh
Odor Odorless
Physical Form Granular
Vapor Pressure 0 hPa
Color Silver-White
Chemical Name or Material Calcium

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H261

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 213-179-5

RTECSNumber : EV8040000

TSCA : Yes

Recommended Storage : Ambient temperatures