Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium granules, redistilled, -16 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number aa4291709
Click to view available options
Quantity:
10 g
50 g
250 g
7440-70-2
Ca
40.08
MFCD00010897 MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]
7440-70-2
Ca
40.08
MFCD00010897 MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Silvery-white granules
Assay Percent Range 99.5% (Metals basis)
Quantity 10 g
Mesh Size -16 Mesh
Odor Odorless
Form Granular
Chemical Name or Material Calcium, Redistilled
Hazard Category H261
Precautionary Statement P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: II
EINECSNumber 213-179-5
RTECSNumber EV8040000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit