Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium hydride, 92% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 4159036

Quantity 500g

 View more versions of this product

Catalog No. AA4159036


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7789-78-8
92% min.
42.1
Calcium hydride
1 To 20mm Granules
CaH2
MFCD00010912

Specifications

7789-78-8
92% min.
Light gray solid
MFCD00010912
Light gray
Calcium hydride
1 To 20mm Granules
CaH2
42.1
1.9g/cm3 at 20°C
500g

Safety and Handling

SDS & Safety Documents

H260 : In contact with water releases flammable gases which may ignite spontaneously.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 232-189-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit