Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
7789-78-8
CaH2
42.09
MFCD00010897
7789-78-8
CaH2
42.09
MFCD00010897
Assay Percent Range 92% min. (Total Ca)
Linear Formula CaH2
Fieser 01,105
Merck Index 15,1674
Solubility Information Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid,soluble only in salt melts
Formula Weight 42.09
Percent Purity ca. 93%
Grade Extra Pure
Chloride 5000ppm max
Density 1.9000g/mL
Packaging Plastic bottle
Solubility Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
Specific Gravity 1.9
Melting Point 675.0°C
Quantity 100g
Chemical Name or Material Calcium hydride, extra pure, 0-2 mm grain size
Show More Show Less

GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes skin irritation.
Causes serious eye irritation.
Reacts violently with water.

GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF ON S

GHS Signal Word: Danger

EINECSNumber : 232-189-2