Alfa Aesar™ Calcium hydrogen phosphate dihydrate, 98% min

Manufacturer: Alfa Aesar™ 40233A1

Quantity 1kg

 View more versions of this product

Catalog No. AA40233A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-77-7
    Linear Formula CaHPO4·2H2O
    Formula Weight 172.09 (136.06 Anhydrous)
    MDL Number MFCD00149621
    Merck Index 14,1692
    Synonym Calcium phosphate, dibasic dihydrate; Calcium orthophosphate dihydrate
    Chemical Name or Material Calcium hydrogen phosphate dihydrate

    Specifications

    Formula Weight 172.09 (136.06 Anhydrous)
    Melting Point 109°C (in -H2O)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Merck Index 14,1692
    Synonym Calcium phosphate, dibasic dihydrate; Calcium orthophosphate dihydrate
    CAS 7789-77-7
    Color White
    Quantity 1kg
    Health Hazard 1 Warning
    Physical Form Powder
    MDL Number MFCD00149621
    Solubility Information Slightly soluble in water; Soluble in dilute hydrochloric, nitric, and acetic acid; Insoluble in alcohol
    TSCA TSCA
    Linear Formula CaHPO4·2H2O
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 2.306g/mL
    Chemical Name or Material Calcium hydrogen phosphate dihydrate
    Assay ≤98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit