Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium hydroxide, 99.995%, (trace metal basis), Acros Organics

Click to view available options
Quantity:
25g
1305-62-0
H2CaO2
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium hydroxide,slaked lime,hydrated lime,calcium dihydroxide,calcium ii hydroxide,calcium oxide, hydrated,kalziumhydroxid,loeschkalk,pickling lime,calciumdihydroxid
6093208
CHEBI:31341
calcium;dihydroxide
[OH-].[OH-].[Ca+2]
1305-62-0
H2CaO2
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium hydroxide,slaked lime,hydrated lime,calcium dihydroxide,calcium ii hydroxide,calcium oxide, hydrated,kalziumhydroxid,loeschkalk,pickling lime,calciumdihydroxid
6093208
CHEBI:31341
calcium;dihydroxide
[OH-].[OH-].[Ca+2]

For ultra trace analysis

Assay Percent Range 54% min. (Ca) (Complexometry)
Linear Formula Ca(OH)2
Merck Index 15,1675
Solubility Information Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol
Formula Weight 74.09
Percent Purity 99.995%
Grade Trace Metal Basis
Solubility Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol
Total Trace Metal Impurities 50ppm max
Boiling Point 2850.0°C
Melting Point 580.0°C
Quantity 25g
Chemical Name or Material Calcium hydroxide
Show More Show Less

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 215-137-3